Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrachloroethylene (Stabilized/Reagent), Fisher Chemical

Catalog Number o45864
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
127-18-4
C2Cl4
165.82
MFCD00000834
CYTYCFOTNPOANT-UHFFFAOYSA-N
tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin
31373
CHEBI:17300
tetrachloroethene
ClC(Cl)=C(Cl)Cl
127-18-4
C2Cl4
165.82
MFCD00000834
CYTYCFOTNPOANT-UHFFFAOYSA-N
tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin
31373
CHEBI:17300
tetrachloroethene
ClC(Cl)=C(Cl)Cl

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

TRUSTED_SUSTAINABILITY
Boiling Range 4°C max.
Decomposition Information >150°C
Melting Point -22°C
Color Colorless
Density 1.625g/cm³
Boiling Point 120°C
Packaging Glass Bottle
Linear Formula CCl2:CCl2
Quantity 4 L
Stabilizer 0.2 to 1%
Vapor Pressure 18mbar at 20°C
Formula Weight 165.83
Grade Reagent
Physical Form Liquid
Chemical Name or Material Tetrachloroethylene, Stabilized
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Suspect cancer hazard. Irritating to eyes and skin. May cause irritation of respiratory tract. May cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:2
Flammability:0
Instability:0
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit