Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrachlorophthalic anhydride, 98%, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
Packaging:
Plastic bottle
117-08-8,118-74-1
C8Cl4O3
285.89
MFCD00005920
AUHHYELHRWCWEZ-UHFFFAOYSA-N
tetrachlorophthalic anhydride,tetrathal,4,5,6,7-tetrachloro-1,3-isobenzofurandione,niagathal,1,3-isobenzofurandione, 4,5,6,7-tetrachloro,phthalic anhydride, tetrachloro,4,5,6,7-tetrachloroisobenzofuran-1,3-dione,1,3-dioxy-4,5,6,7-tetrachloroisobenzofuran,unii-76glw0lbek,ccris 6202
8326
CHEBI:59097
4,5,6,7-tetrachloro-2-benzofuran-1,3-dione
C12=C(C(=C(C(=C1Cl)Cl)Cl)Cl)C(=O)OC2=O
117-08-8,118-74-1
C8Cl4O3
285.89
MFCD00005920
AUHHYELHRWCWEZ-UHFFFAOYSA-N
tetrachlorophthalic anhydride,tetrathal,4,5,6,7-tetrachloro-1,3-isobenzofurandione,niagathal,1,3-isobenzofurandione, 4,5,6,7-tetrachloro,phthalic anhydride, tetrachloro,4,5,6,7-tetrachloroisobenzofuran-1,3-dione,1,3-dioxy-4,5,6,7-tetrachloroisobenzofuran,unii-76glw0lbek,ccris 6202
8326
CHEBI:59097
4,5,6,7-tetrachloro-2-benzofuran-1,3-dione
C12=C(C(=C(C(=C1Cl)Cl)Cl)Cl)C(=O)OC2=O
Insoluble Matter 0.5% max. (10 g in 100 ml 1 M NaOH)
Melting Point 254.0°C to 258.0°C
Packaging Plastic bottle
Boiling Point 349.0°C to 354.0°C
Infrared Spectrum Authentic
Quantity 1kg
Color White
Assay Percent Range 97.5% min. (HPLC)
Beilstein 17, 484
Fieser 15,300
Solubility Information Solubility in water: 0.8 mg/l (21°C). Other solubilities: hydrolysis to acid in water
Formula Weight 285.89
Physical Form Flakes
Percent Purity 98%
Chemical Name or Material Tetrachlorophthalic anhydride, 98%
Show More Show Less

GHS H Statement
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
May cause canc

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not handle until all safety precaut

GHS Signal Word: Danger

EINECSNumber : 204-171-4

RTECSNumber : TI3450000

TSCA : TSCA