Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrachlorophthalic anhydride, 98%, Thermo Scientific Chemicals

Catalog Number ac157792500
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
117-08-8, 118-74-1
C8Cl4O3
285.89
MFCD00005920
AUHHYELHRWCWEZ-UHFFFAOYSA-N
tetrachlorophthalic anhydride, tetrathal, 4,5,6,7-tetrachloro-1,3-isobenzofurandione, niagathal, 1,3-isobenzofurandione, 4,5,6,7-tetrachloro, phthalic anhydride, tetrachloro, 4,5,6,7-tetrachloroisobenzofuran-1,3-dione, 1,3-dioxy-4,5,6,7-tetrachloroisobenzofuran, unii-76glw0lbek, ccris 6202
8326
CHEBI:59097
4,5,6,7-tetrachloro-2-benzofuran-1,3-dione
C12=C(C(=C(C(=C1Cl)Cl)Cl)Cl)C(=O)OC2=O
117-08-8, 118-74-1
C8Cl4O3
285.89
MFCD00005920
AUHHYELHRWCWEZ-UHFFFAOYSA-N
tetrachlorophthalic anhydride, tetrathal, 4,5,6,7-tetrachloro-1,3-isobenzofurandione, niagathal, 1,3-isobenzofurandione, 4,5,6,7-tetrachloro, phthalic anhydride, tetrachloro, 4,5,6,7-tetrachloroisobenzofuran-1,3-dione, 1,3-dioxy-4,5,6,7-tetrachloroisobenzofuran, unii-76glw0lbek, ccris 6202
8326
CHEBI:59097
4,5,6,7-tetrachloro-2-benzofuran-1,3-dione
C12=C(C(=C(C(=C1Cl)Cl)Cl)Cl)C(=O)OC2=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 254.0°C to 258.0°C
Color White
Boiling Point 349.0°C to 354.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Plastic bottle
Quantity 250 g
Beilstein 17, 484
Fieser 15,300
Solubility Information Solubility in water: 0.8 mg/l (21°C). Other solubilities: hydrolysis to acid in water
Formula Weight 285.89
Percent Purity 98%
Insoluble Matter 0.5% max. (10 g in 100 ml 1 M NaOH)
Physical Form Flakes
Chemical Name or Material Tetrachlorophthalic anhydride, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
May cause canc
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not handle until all safety precaut
EINECSNumber 204-171-4
RTECSNumber TI3450000
TSCA TSCA

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit