Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tetracosanoic acid, 99%, ACROS Organics™
Click to view available options
Quantity:
250mg
1g
5g
Packaging:
Glass bottle
Specifications
Color | White |
Quantity | 1g |
Flash Point | >110°C |
Packaging | Glass bottle |
Infrared Spectrum | Authentic |
Melting Point | 84.0°C to 86.0°C |
Boiling Point | 272.0°C (10.0 mmHg) |
Assay Percent Range | 98.5% min. (GC) |
Linear Formula | CH3(CH2)22CO2H |
Beilstein | 02, I, 181 |
Show More |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective cloth
GHS Signal Word: Warning
EINECSNumber : 209-180-7