Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetracosanoic acid, 99%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
5g
Packaging:
Glass bottle
557-59-5
C24H48O2
368.63
MFCD00002810
QZZGJDVWLFXDLK-UHFFFAOYSA-N
lignoceric acid,unii-rk3vcw5y1l,n-tetracosanoic acid,rk3vcw5y1l,n-tetracosanoate,lignozerinsaeure,tetracosansaeure,carnaubic acid,tetracosoic acid,tetracosanic acid
11197
CHEBI:28866
tetracosanoic acid
CCCCCCCCCCCCCCCCCCCCCCCC(=O)O
557-59-5
C24H48O2
368.63
MFCD00002810
QZZGJDVWLFXDLK-UHFFFAOYSA-N
lignoceric acid,unii-rk3vcw5y1l,n-tetracosanoic acid,rk3vcw5y1l,n-tetracosanoate,lignozerinsaeure,tetracosansaeure,carnaubic acid,tetracosoic acid,tetracosanic acid
11197
CHEBI:28866
tetracosanoic acid
CCCCCCCCCCCCCCCCCCCCCCCC(=O)O
Packaging Glass bottle
Color White
Quantity 250mg
Melting Point 84.0°C to 86.0°C
Flash Point >110°C
Boiling Point 272.0°C (10.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Linear Formula CH3(CH2)22CO2H
Beilstein 02, I, 181
Merck Index 15, 5541
Solubility Information Solubility in water: .. Other solubilities: practically insoluble in ethanol, soluble in hot hydrocarbons and hot ketones
Formula Weight 368.63
Physical Form Shiny Crystalline Powder or Flakes
Percent Purity 99%
Chemical Name or Material Tetracosanoic acid
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective cloth

GHS Signal Word: Warning

EINECSNumber : 209-180-7