Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetracyanoethylene, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
100g
Packaging:
Glass bottle
670-54-2
C6N4
128.09
NLDYACGHTUPAQU-UHFFFAOYSA-N
tetracyanoethylene,tetracyanoethene,ethylenetetracarbonitrile,tcne,ethenetetracarbonitrile,ethene, tetracyano,1,1,2,2-ethenetetracarbonitrile,tetrakyanethylen,1,1,2,2-tetracyanoethene,1,1,2,2-tetracyanoethylene
12635
ethene-1,1,2,2-tetracarbonitrile
C(#N)C(=C(C#N)C#N)C#N
670-54-2
C6N4
128.09
NLDYACGHTUPAQU-UHFFFAOYSA-N
tetracyanoethylene,tetracyanoethene,ethylenetetracarbonitrile,tcne,ethenetetracarbonitrile,ethene, tetracyano,1,1,2,2-ethenetetracarbonitrile,tetrakyanethylen,1,1,2,2-tetracyanoethene,1,1,2,2-tetracyanoethylene
12635
ethene-1,1,2,2-tetracarbonitrile
C(#N)C(=C(C#N)C#N)C#N
C-X bond formation reagent, Oxidation reagent
Quantity 100g
Infrared Spectrum Authentic
Boiling Point 223.0°C
Packaging Glass bottle
Color Undesignated
Melting Point 196.0°C to 201.0°C
Assay Percent Range 98%
Linear Formula (NC)2C=C(CN)2
Fieser 01,1133; 02,397; 05,647; 06,567; 13,289
Merck Index 15,934
Solubility Information Solubility in water: hydrolyzes (forming hcn) in water. Other solubilities: slightly soluble in: ether, benzene,, carbon tetrachloride, soluble in: acetone
Formula Weight 128.09
Physical Form Crystalline Powder, Crystals or Chunks
Percent Purity 98%
Chemical Name or Material Tetracyanoethylene
Show More Show Less

GHS P Statement
Fatal in contact with skin.
Fatal if swallowed.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Rinse mouth.
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

Danger