Learn More
Medchemexpress LLC Tetracycline | 60-54-8 | 98.0% | C22H24N2O8 | 50 G
SDP
Supplier:  Medchemexpress LLC HYA010750G
Encompass_Preferred
Tetracycline is a broad-spectrum antibiotic with oral activity, effective against a wide range of bacteria, including gram-positive and gram-negative bacteria, chlamydiae, mycoplasmas, and rickettsiae. It is suitable for research on infections and in animal modeling for genetically engineered disease models.
  • Broad-spectrum oral antibiotic.
  • Effective against gram-positive and gram-negative bacteria, chlamydiae, mycoplasmas, and rickettsiae.
  • Suitable for infection research.
  • Inhibits L-amyloid aggregate formation.
  • Used in animal modeling to create genetically engineered disease models.
  • Can induce tumor regression in mice by inhibiting BCR-ABL expression.
Catalog No. 50-004-32782