Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Tetracycline hydrochloride, 10mg/mL in 100% methanol, sterile-filtered

Click to view available options
Quantity:
50mL
100mL
64-75-5
C22H25ClN2O8
480.898
MFCD00078142
HTXDZWDXSWLLLW-FMZCEJRJSA-N
tetracycline hydrochloride
129628373
(4S,4aS,5aS,6S,12aS)-4-(dimethylamino)-3,6,10,11,12a-pentahydroxy-6-methyl-1,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
CC1(C2CC3C(C(=C(C(=O)C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)C(=O)N)O)N(C)C)O.Cl
64-75-5
C22H25ClN2O8
480.898
MFCD00078142
HTXDZWDXSWLLLW-FMZCEJRJSA-N
tetracycline hydrochloride
129628373
(4S,4aS,5aS,6S,12aS)-4-(dimethylamino)-3,6,10,11,12a-pentahydroxy-6-methyl-1,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
CC1(C2CC3C(C(=C(C(=O)C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)C(=O)N)O)N(C)C)O.Cl
Quantity 100mL
UN Number UN1230
Physical Form Liquid
Chemical Name or Material Tetracycline hydrochloride, 10mg/mL in 100% methanol, sterile-filtered

GHS H Statement
H225-H311-H331-H370-H361
Highly flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Suspected of damaging fertility or the unborn child.

GHS P Statement
P210-P260-P303+P361+P353-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: METHANOL

RTECSNumber : PC1400000

TSCA : TSCA

storageNote1 : -30° to -10°C