Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetradecylbenzene, 98%, Acros Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
1459-10-5
C20H34
274.492
MFCD00015080
JZALLXAUNPOCEU-UHFFFAOYSA-N
1-phenyltetradecane,benzene, tetradecyl,1-tetradecylbenzene,tetradecane, 1-phenyl,myristylbenzene,n-tetradecylbenzene,unii-i0y9j0o95a,phenyl-n-tetradecane,acmc-209cvz,1-phenyltetradecane, analytical standard
15086
tetradecylbenzene
CCCCCCCCCCCCCCC1=CC=CC=C1
1459-10-5
C20H34
274.492
MFCD00015080
JZALLXAUNPOCEU-UHFFFAOYSA-N
1-phenyltetradecane,benzene, tetradecyl,1-tetradecylbenzene,tetradecane, 1-phenyl,myristylbenzene,n-tetradecylbenzene,unii-i0y9j0o95a,phenyl-n-tetradecane,acmc-209cvz,1-phenyltetradecane, analytical standard
15086
tetradecylbenzene
CCCCCCCCCCCCCCC1=CC=CC=C1
Refractive Index 1.4805 to 1.4825 (20°C, 589nm)
Color Colorless to Yellow
Packaging Glass bottle
Quantity 25g
Density 0.8500g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula C6H5(CH2)13CH3
Formula Weight 274.49
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material Tetradecylbenzene, 99%
Show More Show Less

EINECSNumber : 215-950-3

TSCA : TSCA