Alfa Aesar™ Tetraethylammonium borohydride

Manufacturer: Alfa Aesar™ 3257304

Quantity 2g

 View more versions of this product

Catalog No. AA3257304

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 17083-85-1
    Linear Formula (C2H5)4NBH4
    Formula Weight 145.1
    MDL Number MFCD00031600
    UN Number UN3131
    Beilstein 3693695
    Synonym Tetraethylammonium tetrahydridoborate
    Chemical Name or Material Tetraethylammonium borohydride

    Specifications

    Formula Weight 145.1
    Health Hazard 3 GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310-P370+P378i-P402+P404
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    In case of fire: Use for extinction: Fire-extinguishing powder.
    Store in a dry place.
    Store in a closed container.
    Physical Form Powder
    MDL Number MFCD00031600
    Beilstein 3693695
    Synonym Tetraethylammonium tetrahydridoborate
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H261-H314-H318-H302
    In contact with water releases flammable gas.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Color White
    TSCA TSCA
    CAS 17083-85-1
    UN Number UN3131
    DOT Information Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: WATER-REACTIVE SOLID, CORROSIVE, N.O.S.
    Density 0.926g/mL
    Melting Point ≈225°C (dec.)
    Linear Formula (C2H5)4NBH4
    RTECS Number BS5850000
    Odor Odorless
    Quantity 2g
    Sensitivity Moisture Sensitive
    Chemical Name or Material Tetraethylammonium borohydride

    GHS H Statement
    H261-H314-H318-H302
    In contact with water releases flammable gas.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.

    GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310-P370+P378i-P402+P404
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    In case of fire: Use for extinction: Fire-extinguishing powder.
    Store in a dry place.
    Store in a closed container.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit