Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraethylammonium Chloride Hydrate 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
56-34-8
C8H20ClN·xH2O
165.71
MFCD00149992
YMBCJWGVCUEGHA-UHFFFAOYSA-M
tetraethylammonium chloride,tetraethyl ammonium chloride,etamon chloride,tea chloride,teac,n,n,n-triethylethanaminium chloride,tetraethylammoniumchloride,ethanaminium, n,n,n-triethyl-, chloride,tetraethyl-ammonium chloride,unii-8b82z83xnn
5946
CHEBI:78161
tetraethylazanium;chloride
CC[N+](CC)(CC)CC.[Cl-]
56-34-8
C8H20ClN·xH2O
165.71
MFCD00149992
YMBCJWGVCUEGHA-UHFFFAOYSA-M
tetraethylammonium chloride,tetraethyl ammonium chloride,etamon chloride,tea chloride,teac,n,n,n-triethylethanaminium chloride,tetraethylammoniumchloride,ethanaminium, n,n,n-triethyl-, chloride,tetraethyl-ammonium chloride,unii-8b82z83xnn
5946
CHEBI:78161
tetraethylazanium;chloride
CC[N+](CC)(CC)CC.[Cl-]
Phase transfer catalyst
Loss on Drying 10% max.
Quantity 100g
Packaging Glass bottle
Infrared Spectrum Authentic
Color White
Assay Percent Range 99%
Linear Formula (C2H5)4NCl·xH2O
Merck Index 15, 9345
Solubility Information Solubility in water: in water: 1410g/L (25°C). Other solubilities: freely soluble in alcohol, chloroform and acetone, slightly soluble in benzene and ether
Formula Weight 165.71
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material Tetraethylammonium chloride hydrate, 99%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 200-267-5

RTECSNumber : BS6125000

TSCA : TSCA