Alfa Aesar™ Tetraethylammonium chloride monohydrate, 98+%

Phase transfer catalyst

Manufacturer: Alfa Aesar™ 3081830

Quantity 250g

 View more versions of this product

Catalog No. AA3081830

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 68696-18-4
    Formula Weight 183.72 (165.71 Anhydrous)
    MDL Number MFCD00149992
    Beilstein 3563247
    Merck Index 14,9200
    Chemical Name or Material Tetraethylammonium chloride monohydrate

    Specifications

    Formula Weight 183.72 (165.71 Anhydrous)
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number BS6125000
    Physical Form Crystalline
    MDL Number MFCD00149992
    Beilstein 3563247
    Quantity 250g
    Density 1.0801g/mL
    Merck Index 14,9200
    Health Hazard 1 Warning
    TSCA TSCA
    CAS 68696-18-4
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Melting Point ≈110°C
    Chemical Name or Material Tetraethylammonium chloride monohydrate
    Assay ≥98%

    GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit