Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraethylammonium hydroxide, 20 wt.% in water, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
5L
Packaging:
Plastic bottle
77-98-5,7732-18-5
C8H21NO
147.26
MFCD00009024
LRGJRHZIDJQFCL-UHFFFAOYSA-M
tetraethylammonium hydroxide,tetraethyl ammonium hydroxide,unii-ra8vu41b1f,ammonium, tetraethyl-, hydroxide,tetraethylazanium hydroxide,ethanaminium, n,n,n-triethyl-, hydroxide,ra8vu41b1f,ethanaminium, n,n,n-triethyl-, hydroxide 1:1,tetraethylammonium hydroxide solution in water,n,n,n-triethylethanaminium
6509
tetraethylazanium;hydroxide
CC[N+](CC)(CC)CC.[OH-]
77-98-5,7732-18-5
C8H21NO
147.26
MFCD00009024
LRGJRHZIDJQFCL-UHFFFAOYSA-M
tetraethylammonium hydroxide,tetraethyl ammonium hydroxide,unii-ra8vu41b1f,ammonium, tetraethyl-, hydroxide,tetraethylazanium hydroxide,ethanaminium, n,n,n-triethyl-, hydroxide,ra8vu41b1f,ethanaminium, n,n,n-triethyl-, hydroxide 1:1,tetraethylammonium hydroxide solution in water,n,n,n-triethylethanaminium
6509
tetraethylazanium;hydroxide
CC[N+](CC)(CC)CC.[OH-]
Phase transfer catalyst
Refractive Index 1.402
Density 1.0200g/mL
Quantity 5L
Color Colorless to Yellow
Packaging Plastic bottle
Specific Gravity 1.02
Boiling Point 102.0°C
Linear Formula (C2H5)4NOH
Merck Index 15, 9346
Solubility Information Solubility in water: soluble.
Formula Weight 147.26
Physical Form Liquid
Percent Purity ≥20%
Chemical Name or Material Tetraethylammonium hydroxide, 20 wt.% in Water
Show More Show Less

GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Causes damage to organs if swallowed.
Causes damage to organs through prolonged or repeated exposure in contact with s

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 201-073-3

RTECSNumber : KH3150000

TSCA : TSCA