Learn More
Tetraethylammonium hydroxide, 20 wt.% in water, Thermo Scientific Chemicals
$99.00 - $829.00
Specifications
Chemical Name or Material | Tetraethylammonium hydroxide |
---|---|
Name Note | 20 wt.% in Water |
CAS | 77-98-5, 7732-18-5 |
Color | Colorless to Yellow |
Density | 1.0200g/mL |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
AC176072500
|
Thermo Scientific Chemicals
176072500 |
250 mL | Plastic bottle |
Each for $99.00
Order in Bulk
Request bulk or custom formats |
|
||||
AC176070010
|
Thermo Scientific Chemicals
176070010 |
1 L | Plastic bottle |
Each for $260.00
Order in Bulk
Request bulk or custom formats |
|
||||
AC176070050
|
Thermo Scientific Chemicals
176070050 |
5 L | Plastic bottle |
Each for $829.00
Order in Bulk
Request bulk or custom formats |
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalystSpecifications
Tetraethylammonium hydroxide | |
77-98-5, 7732-18-5 | |
1.0200g/mL | |
Plastic bottle | |
(C2H5)4NOH | |
MFCD00009024 | |
1.02 | |
Solubility in water: soluble. | |
[OH-].CC[N+](CC)(CC)CC | |
147.26 | |
147.26 | |
Liquid |
20 wt.% in Water | |
Colorless to Yellow | |
102.0°C | |
C8H21NO | |
1.402 | |
15, 9346 | |
tetraethylammonium hydroxide, tetraethyl ammonium hydroxide, unii-ra8vu41b1f, ammonium, tetraethyl-, hydroxide, tetraethylazanium hydroxide, ethanaminium, n,n,n-triethyl-, hydroxide, ra8vu41b1f, ethanaminium, n,n,n-triethyl-, hydroxide 1:1, tetraethylammonium hydroxide solution in water, n,n,n-triethylethanaminium | |
LRGJRHZIDJQFCL-UHFFFAOYSA-M | |
tetraethylazanium;hydroxide | |
6509 | |
≥20% |
Safety and Handling
GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Causes damage to organs if swallowed.
Causes damage to organs through prolonged or repeated exposure in contact with s
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 201-073-3
RTECSNumber : KH3150000
TSCA : TSCA
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.