Alfa Aesar™ Tetraethylammonium hydroxide, 25% w/w in methanol

Manufacturer: Alfa Aesar™ 1632809

Quantity 10g

 View more versions of this product

Catalog No. AA1632809

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 77-98-5
    Formula Weight 147.26
    MDL Number MFCD00009024
    UN Number UN2924
    Beilstein 3912961
    Merck Index 14,9201
    Chemical Name or Material Tetraethylammonium hydroxide

    Specifications

    Formula Weight 147.26
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Specific treatment is urgent (see ... on this label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H224-H300-H310-H330-H370-H314-H318
    Extremely flammable liquid and vapor.
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Causes damage to organs.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    MDL Number MFCD00009024
    Refractive Index 1.380
    Odor Like alcohol
    Density 0.88g/mL
    Physical Form Liquid
    UN Number UN2924
    Beilstein 3912961
    Merck Index 14,9201
    Flash Point 16° (60°F)
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
    TSCA TSCA
    CAS 77-98-5
    Health Hazard 1 Danger
    Quantity 10g
    Sensitivity Air Sensitive
    Concentration 25% w/w in methanol
    Chemical Name or Material Tetraethylammonium hydroxide

    GHS H Statement
    H224-H300-H310-H330-H370-H314-H318
    Extremely flammable liquid and vapor.
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Causes damage to organs.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P210-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Specific treatment is urgent (see ... on this label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit