Alfa Aesar™ Tetraethylammonium nitrate, 35% w/w aq. soln.

Manufacturer: Alfa Aesar™ 41791AD

Quantity 50mL

 View more versions of this product

Catalog No. AA41791AD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1941-26-0
    Formula Weight 192.26
    MDL Number MFCD00041978
    UN Number UN3139
    Beilstein 3918466
    Chemical Name or Material Tetraethylammonium nitrate

    Specifications

    Formula Weight 192.26
    Health Hazard 3 GHS P Statement
    P221-P210-P220-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Keep/Store away from clothing/combustible materials.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    UN Number UN3139
    Beilstein 3918466
    Quantity 50mL
    DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: OXIDIZING LIQUID, N.O.S.
    Density 1.029g/mL
    Physical Form Liquid
    MDL Number MFCD00041978
    Health Hazard 1 Danger
    Boiling Point >100°C
    TSCA TSCA
    CAS 1941-26-0
    Health Hazard 2 GHS H Statement
    H272-H315-H319-H335
    May intensify fire
    oxidizer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Concentration 35% w/w aq. soln.
    Chemical Name or Material Tetraethylammonium nitrate

    GHS H Statement
    H272-H315-H319-H335
    May intensify fire
    oxidizer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P221-P210-P220-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Keep/Store away from clothing/combustible materials.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit