Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tetraethylbutylene-1,4-diphosphonate, 98%, Acros Organics
Click to view available options
Quantity:
5g
Packaging:
Glass bottle
Description
For C-C Bond Formation (Olefination)
Specifications
Packaging | Glass bottle |
Color | Colorless |
Quantity | 5g |
Infrared Spectrum | Authentic |
Refractive Index | 1.4465 to 1.4485 (20°C, 589nm) |
Assay Percent Range | 98% |
Formula Weight | 330.3 |
Physical Form | Liquid |
Percent Purity | ≥97.5% |
Chemical Name or Material | Tetraethylbutylene-1, 4-diphosphonate, 98% |