Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraethylene glycol, 99.5%, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
10L
Packaging:
Glass Bottle
Plastic drum
112-60-7
C8H18O5
194.23
UWHCKJMYHZGTIT-UHFFFAOYSA-N
tetraethylene glycol,tetraglycol,2,2'-oxybis ethane-2,1-diyl bis oxy diethanol,hi-dry,carbitol, diethyl,3,6,9-trioxaundecane-1,11-diol,bis 2-2-hydroxyethoxy ethyl ether,ethanol, 2,2'-oxybis 2,1-ethanediyloxy bis,tetra ethylene glycol,hsdb 843
8200
CHEBI:44920
2-[2-[2-(2-hydroxyethoxy)ethoxy]ethoxy]ethanol
C(COCCOCCOCCO)O
112-60-7
C8H18O5
194.23
UWHCKJMYHZGTIT-UHFFFAOYSA-N
tetraethylene glycol,tetraglycol,2,2'-oxybis ethane-2,1-diyl bis oxy diethanol,hi-dry,carbitol, diethyl,3,6,9-trioxaundecane-1,11-diol,bis 2-2-hydroxyethoxy ethyl ether,ethanol, 2,2'-oxybis 2,1-ethanediyloxy bis,tetra ethylene glycol,hsdb 843
8200
CHEBI:44920
2-[2-[2-(2-hydroxyethoxy)ethoxy]ethoxy]ethanol
C(COCCOCCOCCO)O
Viscosity 44.9 mPa.s (25°C)
Density 1.1200g/mL
Flash Point 176°C
Specific Gravity 1.12
Refractive Index 1.4580 to 1.4600
Packaging Plastic drum
Infrared Spectrum Authentic
Boiling Point 324.0°C to 330.0°C
Melting Point -4.1°C
Color Yellow
Quantity 10L
Assay Percent Range 99.5%
Linear Formula O(CH2CH2OCH2CH2OH)2
Beilstein 01,468
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in benzene and toluene, practically insoluble in diethylether
Formula Weight 194.23
Physical Form Liquid
Percent Purity 99.5%
Chemical Name or Material Tetraethylene glycol
Show More Show Less