Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraethylphosphonium Bromide 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
4317-07-1
C8H20BrP
227.13
MFCD00054230
LIXPXSXEKKHIRR-UHFFFAOYSA-M
9859378
tetraethylphosphanium;bromide
CC[P+](CC)(CC)CC.[Br-]
4317-07-1
C8H20BrP
227.13
MFCD00054230
LIXPXSXEKKHIRR-UHFFFAOYSA-M
9859378
tetraethylphosphanium;bromide
CC[P+](CC)(CC)CC.[Br-]
Color White
Quantity 25g
Formula Weight 227.13
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (T)
Chemical Name or Material Tetraethylphosphonium Bromide