Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraethylpropylene-1,3-diphosphonate, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
22401-25-8
C11H26O6P2
316.27
MFCD00048543
LJQJJGQRJQSYQN-UHFFFAOYSA-N
phosphonic acid, trimethylenedi-, tetraethyl ester,phosphonic acid, 1,3-propanediylbis-, tetraethyl ester,tetraethyl propylenediphosphonate,tetraethyltrimethylenediphosphonate,1,3-bis diethoxyphosphinyl propane,1,3-bis diethoxyphosphoryl propane,tetraethyl propane-1,3-diphosphonate,tetraethyl 1,3-propylene bisphosphonate,propanediyl-bis-phosphonic acid tetraethyl ester,propane-1,3-diphosphonic acid, tetraethyl ester
519945
1,3-bis(diethoxyphosphoryl)propane
CCOP(=O)(CCCP(=O)(OCC)OCC)OCC
22401-25-8
C11H26O6P2
316.27
MFCD00048543
LJQJJGQRJQSYQN-UHFFFAOYSA-N
phosphonic acid, trimethylenedi-, tetraethyl ester,phosphonic acid, 1,3-propanediylbis-, tetraethyl ester,tetraethyl propylenediphosphonate,tetraethyltrimethylenediphosphonate,1,3-bis diethoxyphosphinyl propane,1,3-bis diethoxyphosphoryl propane,tetraethyl propane-1,3-diphosphonate,tetraethyl 1,3-propylene bisphosphonate,propanediyl-bis-phosphonic acid tetraethyl ester,propane-1,3-diphosphonic acid, tetraethyl ester
519945
1,3-bis(diethoxyphosphoryl)propane
CCOP(=O)(CCCP(=O)(OCC)OCC)OCC
For C-C Bond Formation (Olefination)
Boiling Point 206.0°C to 227.0°C (5.0mmHg)
Color Colorless
Quantity 5g
Packaging Glass bottle
Infrared Spectrum Authentic
Refractive Index 1.4450 to 1.4470
Assay Percent Range 96% min. (GC)
Formula Weight 316.27
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Tetraethylpropylene-1, 3-diphosphonate, 97%
Show More Show Less