Alfa Aesar™ Tetraethyltin, 98%

Manufacturer: Alfa Aesar™ 7114403

Quantity 1g

 View more versions of this product

Catalog No. AA7114403

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 597-64-8
    Molecular Formula (C2H5)4Sn
    Formula Weight 234.94
    MDL Number MFCD00009020
    UN Number UN3384
    Synonym Tetraethylstannane
    Chemical Name or Material Tetraethyltin

    Specifications

    Formula Weight 234.94
    Health Hazard 3 GHS P Statement
    P273-P303+P361+P353-P309-P310-P501a
    Avoid release to the environment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Physical Form Liquid
    CAS 597-64-8
    Health Hazard 2 GHS H Statement
    H300-H310-H330-H226
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Flammable liquid and vapor.
    Density 1.187g/mL
    Boiling Point 181°C
    Melting Point −112°C
    Molecular Formula (C2H5)4Sn
    MDL Number MFCD00009020
    UN Number UN3384
    Solubility Information Insoluble in water; Soluble in alcohol and ether
    Flash Point 53°C (128°F)
    Refractive Index 1.4730
    Quantity 1g
    Synonym Tetraethylstannane
    Health Hazard 1 Danger
    Odor Characteristic
    DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC BY INHALATION LIQUID, FLAMMABLE, N.O.S.
    RTECS Number WH8625000
    Chemical Name or Material Tetraethyltin
    Assay 98%

    GHS H Statement
    H300-H310-H330-H226
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Flammable liquid and vapor.

    GHS P Statement
    P273-P303+P361+P353-P309-P310-P501a
    Avoid release to the environment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit