Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Tetrafluorophthalonitrile, 96%

Click to view available options
Quantity:
1g
5g
1835-65-0
C8F4N2
200.096
MFCD00001774
OFLRJMBSWDXSPG-UHFFFAOYSA-N
tetrafluorophthalonitrile,3,4,5,6-tetrafluorophthalonitrile,1,2-dicyano-3,4,5,6-tetrafluorobenzene,1,2-benzenedicarbonitrile, 3,4,5,6-tetrafluoro,1,2-benzenedicarbonitrile, 3,4,5,6-tetrafluoro-;3,4,5,6-tetrafluorobenzene-1,2-dicarbonitrile,pubchem4564,acmc-209ekr,ksc496c0b,tetrafluorophthalonitrile,3,4,5,6-tetrafluorophthalonitrile #
74600
3,4,5,6-tetrafluorobenzene-1,2-dicarbonitrile
C(#N)C1=C(C(=C(C(=C1F)F)F)F)C#N
1835-65-0
C8F4N2
200.096
MFCD00001774
OFLRJMBSWDXSPG-UHFFFAOYSA-N
tetrafluorophthalonitrile,3,4,5,6-tetrafluorophthalonitrile,1,2-dicyano-3,4,5,6-tetrafluorobenzene,1,2-benzenedicarbonitrile, 3,4,5,6-tetrafluoro,1,2-benzenedicarbonitrile, 3,4,5,6-tetrafluoro-;3,4,5,6-tetrafluorobenzene-1,2-dicarbonitrile,pubchem4564,acmc-209ekr,ksc496c0b,tetrafluorophthalonitrile,3,4,5,6-tetrafluorophthalonitrile #
74600
3,4,5,6-tetrafluorobenzene-1,2-dicarbonitrile
C(#N)C1=C(C(=C(C(=C1F)F)F)F)C#N
Melting Point 82°C to 86°C
Quantity 5g
UN Number UN3439
Formula Weight 200.09
Percent Purity 96%
Chemical Name or Material Tetrafluorophthalonitrile

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.