Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraformylhydrazine, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass Bottle
52023-52-6
C4H4N2O4
144.086
MFCD08059355
NODMXZGWWULGLE-UHFFFAOYSA-N
tetraformylhydrazine,hydrazinetetracarboxaldehyde,n,n',n'-triformylformohydrazide,1,1,2,2-hydrazinetetracarbox aldehyde
11217319
N-(diformylamino)-N-formylformamide
C(=O)N(C=O)N(C=O)C=O
52023-52-6
C4H4N2O4
144.086
MFCD08059355
NODMXZGWWULGLE-UHFFFAOYSA-N
tetraformylhydrazine,hydrazinetetracarboxaldehyde,n,n',n'-triformylformohydrazide,1,1,2,2-hydrazinetetracarbox aldehyde
11217319
N-(diformylamino)-N-formylformamide
C(=O)N(C=O)N(C=O)C=O
Packaging Glass Bottle
Color White
Quantity 5g
Infrared Spectrum Authentic
Elemental Analysis 97.5% min.
Melting Point 180.0°C to 183.0°C
Assay Percent Range 98%
Linear Formula [N(COH)2]2
Formula Weight 144.09
Physical Form Powder
Percent Purity 98%
Chemical Name or Material Tetraformylhydrazine
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.