Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraheptylammonium chloride, 95%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
Packaging:
Glass bottle
10247-90-2
C28H60ClN
446.245
MFCD00041983
VMJQVRWCDVLJSI-UHFFFAOYSA-M
tetraheptylammonium chloride,1-heptanaminium, n,n,n-triheptyl-, chloride 1:1,1-heptanaminium, n,n,n-triheptyl-, chloride,n,n,n-triheptylheptan-1-aminium chloride,tetraheptylazanium chloride,tetra-n-heptylammonium cloride,1-heptanaminium,n,n,n-triheptyl-, chloride 1:1
82489
tetraheptylazanium;chloride
CCCCCCC[N+](CCCCCCC)(CCCCCCC)CCCCCCC.[Cl-]
10247-90-2
C28H60ClN
446.245
MFCD00041983
VMJQVRWCDVLJSI-UHFFFAOYSA-M
tetraheptylammonium chloride,1-heptanaminium, n,n,n-triheptyl-, chloride 1:1,1-heptanaminium, n,n,n-triheptyl-, chloride,n,n,n-triheptylheptan-1-aminium chloride,tetraheptylazanium chloride,tetra-n-heptylammonium cloride,1-heptanaminium,n,n,n-triheptyl-, chloride 1:1
82489
tetraheptylazanium;chloride
CCCCCCC[N+](CCCCCCC)(CCCCCCC)CCCCCCC.[Cl-]
Phase transfer catalyst
Packaging Glass bottle
Refractive Index 1.4690 to 1.4710 (20°C, 589nm)
Quantity 25g
Infrared Spectrum Authentic
Color Yellow
Assay Percent Range 95%
Formula Weight 446.24
Physical Form Viscous Liquid
Percent Purity ≥94%
Chemical Name or Material Tetraheptylammonium chloride, 95%

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 233-583-7

TSCA : TSCA