Alfa Aesar™ Tetrahydro-2,2-dimethyl-4H-pyran-4-one, 95%

Manufacturer: Alfa Aesar™ H27774MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH27774MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1194-16-7
    Molecular Formula C7H12O2
    Formula Weight 128.17
    MDL Number MFCD01549337
    Synonym 2,2-Dimethyltetrahydropyran-4-one
    Chemical Name or Material Tetrahydro-2,2-dimethyl-4H-pyran-4-one

    Specifications

    Formula Weight 128.17
    Molecular Formula C7H12O2
    MDL Number MFCD01549337
    CAS 1194-16-7
    Synonym 2,2-Dimethyltetrahydropyran-4-one
    Quantity 250mg
    Recommended Storage 2° to 8°C
    Boiling Point 60° to 64°C (8mmHg)
    Chemical Name or Material Tetrahydro-2,2-dimethyl-4H-pyran-4-one
    Assay 95%

    Description

    Applications
    It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

    Solubility
    Slightly soluble in water.

    Notes
    Store in cool dry conditions. Incompatible with oxidizing agents.

    2° to 8°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit