Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydro-2H-pyran-2-ol, 90%, cyclized form of 5-Hydroxypentanal, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
694-54-2
C5H10O2
102.13
MFCD02683093
CELWCAITJAEQNL-UHFFFAOYSA-N
2-hydroxytetrahydropyran,tetrahydro-2h-pyran-2-ol,oxanol,2h-pyran-2-ol, tetrahydro,2-tetrahydropyranol,tetrahydropyran-2-ol,tetrahydro-2-hydroxy-2h-pyran,tetrahydro-2-pyranol,tetrahydropyranyl alcohol,2h-pyran-2-ol,tetrahydro
136505
oxan-2-ol
C1CCOC(C1)O
694-54-2
C5H10O2
102.13
MFCD02683093
CELWCAITJAEQNL-UHFFFAOYSA-N
2-hydroxytetrahydropyran,tetrahydro-2h-pyran-2-ol,oxanol,2h-pyran-2-ol, tetrahydro,2-tetrahydropyranol,tetrahydropyran-2-ol,tetrahydro-2-hydroxy-2h-pyran,tetrahydro-2-pyranol,tetrahydropyranyl alcohol,2h-pyran-2-ol,tetrahydro
136505
oxan-2-ol
C1CCOC(C1)O
Color Colorless
Quantity 25g
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 90%
Formula Weight 102.13
Physical Form Liquid
Percent Purity 90%
Chemical Name or Material Tetrahydro-2H-pyran-2-ol

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning