Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Pyrazolidinone hydrochloride, 98%, Acros Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
1752-88-1
C3H6N2O·HCl
122.55
HAYWTXQRYXYRAF-UHFFFAOYSA-N
3-pyrazolidinone hydrochloride,pyrazolidin-3-one hydrochloride,4,5-dihydro-1h-pyrazol-3-ol hydrochloride,3-pyrazolidinone, monohydrochloride,tetrahydro-3h-pyrazol-3-one hydrochloride,3-pyrazolidinone hcl,3-pyrazolidone hydrochloride,ksc493k0l,pyrazolidin-3-on-hydrochloride,pyrazolidin-3-one hydrogen chloride
2733712
pyrazolidin-3-one;hydrochloride
C1CNNC1=O.Cl
1752-88-1
C3H6N2O·HCl
122.55
HAYWTXQRYXYRAF-UHFFFAOYSA-N
3-pyrazolidinone hydrochloride,pyrazolidin-3-one hydrochloride,4,5-dihydro-1h-pyrazol-3-ol hydrochloride,3-pyrazolidinone, monohydrochloride,tetrahydro-3h-pyrazol-3-one hydrochloride,3-pyrazolidinone hcl,3-pyrazolidone hydrochloride,ksc493k0l,pyrazolidin-3-on-hydrochloride,pyrazolidin-3-one hydrogen chloride
2733712
pyrazolidin-3-one;hydrochloride
C1CNNC1=O.Cl
Quantity 5g
Infrared Spectrum Authentic
Color White to Yellow
Packaging Glass bottle
Assay Percent Range 98%
Solubility Information Solubility in water: soluble
Formula Weight 122.55
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 3-Pyrazolidinone hydrochloride

GHS Signal Word: Warning