Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydro-4H-pyran-4-ol, Acros Organics

Click to view available options
Quantity:
1g
5g
2081-44-9
C5H10O2
102.13
LMYJGUNNJIDROI-UHFFFAOYSA-N
tetrahydro-2h-pyran-4-ol,tetrahydro-4-pyranol,4-hydroxytetrahydropyran,tetrahydro-4h-pyran-4-ol,tetrahydro-pyran-4-ol,2h-pyran-4-ol, tetrahydro,4-hydroxytetrahydropyan,tetrahydropyran-4-ol,4-hydroxytetrahydro-4h-pyran,4-hydroxytetrahydro-2h-pyran
74956
oxan-4-ol
C1COCCC1O
2081-44-9
C5H10O2
102.13
LMYJGUNNJIDROI-UHFFFAOYSA-N
tetrahydro-2h-pyran-4-ol,tetrahydro-4-pyranol,4-hydroxytetrahydropyran,tetrahydro-4h-pyran-4-ol,tetrahydro-pyran-4-ol,2h-pyran-4-ol, tetrahydro,4-hydroxytetrahydropyan,tetrahydropyran-4-ol,4-hydroxytetrahydro-4h-pyran,4-hydroxytetrahydro-2h-pyran
74956
oxan-4-ol
C1COCCC1O
Quantity 5g
Boiling Point 87.0°C (15.0 mmHg)
Refractive Index 1.46
Flash Point 190°C
Specific Gravity 1.07
Density 1.0700g/mL
Beilstein 17,II,106
Formula Weight 102.13
Chemical Name or Material Tetrahydro-4H-pyran-4-ol

GHS Signal Word: Warning

EINECSNumber : 218-210-8