Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydro-4H-pyran-4-one, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
29943-42-8
C5H8O2
100.12
JMJRYTGVHCAYCT-UHFFFAOYSA-N
tetrahydro-4h-pyran-4-one,4-oxotetrahydropyran,tetrahydropyran-4-one,dihydro-2h-pyran-4 3h-one,4h-pyran-4-one, tetrahydro,4-thp,tetrahydro-2h-pyran-4-one,tetrahydro-pyran-4-one,4-oxotetrahydopyran,tetrahydro-.gamma.-pyrone
121599
oxan-4-one
C1COCCC1=O
29943-42-8
C5H8O2
100.12
JMJRYTGVHCAYCT-UHFFFAOYSA-N
tetrahydro-4h-pyran-4-one,4-oxotetrahydropyran,tetrahydropyran-4-one,dihydro-2h-pyran-4 3h-one,4h-pyran-4-one, tetrahydro,4-thp,tetrahydro-2h-pyran-4-one,tetrahydro-pyran-4-one,4-oxotetrahydopyran,tetrahydro-.gamma.-pyrone
121599
oxan-4-one
C1COCCC1=O
Specific Gravity 1.08
Refractive Index 1.4510 to 1.4530
Packaging Glass bottle
Infrared Spectrum Authentic
Flash Point 56°C
Boiling Point 166.0°C
Color Colorless to Yellow
Quantity 25g
Density 1.0800g/mL
Assay Percent Range 99%
Beilstein 17,I,131
Solubility Information Solubility in water: miscible
Formula Weight 100.12
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Tetrahydro-4H-pyran-4-one
Show More Show Less

GHS H Statement
Flammable liquid and vapour.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.

GHS Signal Word: Warning

EINECSNumber : 249-967-2