Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Tetrahydrofuran-3-one, 95%

Click to view available options
Quantity:
1g
5g
22929-52-8
C4H6O2
86.09
MFCD07778393
JLPJFSCQKHRSQR-UHFFFAOYSA-N
3-oxotetrahydrofuran,dihydrofuran-3 2h-one,dihydro-3 2h-furanone,dihydrofuran-3-one,dihydro-furan-3-one,tetrahydrofuran-3-one,3 2h-furanone, dihydro,3-oxo-1,4-epoxybutane,3 2h-furanone, dihydro,tetrahydrofuran-3-yloxy
529392
oxolan-3-one
C1COCC1=O
22929-52-8
C4H6O2
86.09
MFCD07778393
JLPJFSCQKHRSQR-UHFFFAOYSA-N
3-oxotetrahydrofuran,dihydrofuran-3 2h-one,dihydro-3 2h-furanone,dihydrofuran-3-one,dihydro-furan-3-one,tetrahydrofuran-3-one,3 2h-furanone, dihydro,3-oxo-1,4-epoxybutane,3 2h-furanone, dihydro,tetrahydrofuran-3-yloxy
529392
oxolan-3-one
C1COCC1=O
Quantity 5g
Beilstein 105200
Formula Weight 86.09
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material Tetrahydrofuran-3-one

storageNote1 : 2° to 8°C