Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydrofuran, 99.5%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™, ACROS Organics™

Click to view available options
Quantity:
100mL
500mL
1L
2.5L
109-99-9,128-37-0
C4H8O
72.107
MFCD00005356
WYURNTSHIVDZCO-UHFFFAOYSA-N
tetrahydrofuran,furan, tetrahydro,butylene oxide,furanidine,hydrofuran,tetramethylene oxide,oxacyclopentane,1,4-epoxybutane,tetrahydrofuranne,tetraidrofurano
8028
CHEBI:26911
oxolane
C1CCOC1
109-99-9,128-37-0
C4H8O
72.107
MFCD00005356
WYURNTSHIVDZCO-UHFFFAOYSA-N
tetrahydrofuran,furan, tetrahydro,butylene oxide,furanidine,hydrofuran,tetramethylene oxide,oxacyclopentane,1,4-epoxybutane,tetrahydrofuranne,tetraidrofurano
8028
CHEBI:26911
oxolane
C1CCOC1
Assay Percent Range 99.5%
Beilstein 17, 10
Fieser 01,1140; 02,398; 03,278; 05,649; 06,570
Merck Index 15, 9356
Solubility Information Solubility in water: miscible. Other solubilities: soluble in methanol, ethanol, 2-propanol, dmf,, methylene chloride and acetone
Formula Weight 72.11
Physical Form Liquid
Percent Purity 99.5%
Grade Extra Dry over Molecular Sieve
Density 0.8800g/mL
Refractive Index 1.407
Solubility Solubility in water: miscible. Other solubilities: soluble in methanol, ethanol, 2-propanol, dmf,, methylene chloride and acetone
Specific Gravity 0.88
Stabilizer 100 to 250ppm BHT
Water 0.005% Max. (Coulometric)
Flash Point −21°C
Viscosity 0.55 mPa.s (20°C)
Boiling Point 66.0°C
Color Colorless
Melting Point -108.4°C
Quantity 500mL
Chemical Name or Material Tetrahydrofuran, Water <50 ppm, stabilized
Show More Show Less

GHS P Statement May cause respiratory irritation. Causes serious eye irritation. Highly flammable liquid and vapor. May form explosive peroxides. Suspected of causing cancer.

GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. Avoid breathing dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF ON SKIN: Wash with plenty of soap and water. Keep container tightly closed.

Danger