Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydrofuran, 99.9%, extra pure, anhydrous, stabilized with BHT, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
2.5L
25L
Packaging:
Glass bottle
Metal drum
109-99-9, 128-37-0
C4H8O
72.107
MFCD00005356
WYURNTSHIVDZCO-UHFFFAOYSA-N
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
8028
CHEBI:26911
oxolane
C1CCOC1
109-99-9, 128-37-0
C4H8O
72.107
MFCD00005356
WYURNTSHIVDZCO-UHFFFAOYSA-N
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
8028
CHEBI:26911
oxolane
C1CCOC1

Tetrahydrofuran, >-99.85%, C4H8O, CAS Number-109-99-9, 128-37-0, tetraidrofurano, furanidine, tetrahydrofuranne, butylene oxide, tetrahydrofuran, furan, tetrahydro, oxacyclopentane, hydrofuran, 1,4-epoxybutane, tetramethylene oxide, 1L, 99.9%, 17, 10, CHEBI:26911, Undesignated, 0.8800g/mL

Tetrahydrofuran, >-99.85%, C4H8O, CAS Number-109-99-9, 128-37-0, tetraidrofurano, furanidine, tetrahydrofuranne, butylene oxide, tetrahydrofuran, furan, tetrahydro, oxacyclopentane, hydrofuran, 1,4-epoxybutane, tetramethylene oxide, 1L, 99.9%, 17, 10, CHEBI:26911, Undesignated, 0.8800g/mL

Assay Percent Range 99.9%
Beilstein 17, 10
Fieser 01,1140; 02,398; 03,278; 05,649; 06,570
Merck Index 15, 9356
Formula Weight 72.11
Physical Form Liquid
Percent Purity ≥99.85%
Grade Extra Pure
Density 0.8800g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Peroxides 0.003% max. (as H2O2)
Refractive Index 1.4060 to 1.4080 (20°C, 589nm)
Stabilizer 0.025 to 0.04% BHT
Water 0.006% max. (Coulometric)
Color Undesignated
Quantity 250mL
Chemical Name or Material Tetrahydrofuran, Anhydrous, Stabilized
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
May form explosive peroxides.

GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Keep container tightly closed.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger