Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydrofuran, 99+%, extra pure, stabilized with BHT, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
10L
25L
Packaging:
Glass Bottle
Metal Drum
Plastic drum
109-99-9,128-37-0
C4H8O
72.107
MFCD00005356
WYURNTSHIVDZCO-UHFFFAOYSA-N
tetrahydrofuran,furan, tetrahydro,butylene oxide,furanidine,hydrofuran,tetramethylene oxide,oxacyclopentane,1,4-epoxybutane,tetrahydrofuranne,tetraidrofurano
8028
CHEBI:26911
oxolane
C1CCOC1
109-99-9,128-37-0
C4H8O
72.107
MFCD00005356
WYURNTSHIVDZCO-UHFFFAOYSA-N
tetrahydrofuran,furan, tetrahydro,butylene oxide,furanidine,hydrofuran,tetramethylene oxide,oxacyclopentane,1,4-epoxybutane,tetrahydrofuranne,tetraidrofurano
8028
CHEBI:26911
oxolane
C1CCOC1
Assay Percent Range 99+%
Beilstein 17, 10
Fieser 01,1140; 02,398; 03,278; 05,649; 06,570
Merck Index 15, 9356
Solubility Information Solubility in water: miscible. Other solubilities: soluble in methanol, ethanol, 2-propanol, dmf,, methylene chloride and acetone
Formula Weight 72.11
Physical Form Liquid
Percent Purity ≥99%
Grade Extra Pure
Density 0.8800g/mL
Packaging Glass Bottle
Peroxides 0.015% max. (as H2O2)
Refractive Index 1.407
Residue after Evaporation 0.005% max.
Specific Gravity 0.88
Stabilizer 75 to 400ppm BHT
Water 0.05% max. (Coulometric)
Flash Point −21°C
Viscosity 0.55 mPa.s (20°C)
Boiling Point 66.0°C
Color Undesignated
Melting Point -108.4°C
Quantity 1L
Chemical Name or Material Tetrahydrofuran
Show More Show Less

GHS P Statement May cause respiratory irritation. Causes serious eye irritation. Highly flammable liquid and vapor. May form explosive peroxides. Suspected of causing cancer.

GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. Avoid breathing dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF ON SKIN: Wash with plenty of soap and water. Keep container tightly closed.

Danger