Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydrofuran, Anhydrous, 99.9%, Spectrum™

Click to view available options
Quantity:
100 mL
1 L
Packaging:
Amber Glass Bottle
109-99-9
C4H8O
72.11
WYURNTSHIVDZCO-UHFFFAOYSA-N
oxolane
C1CCOC1
109-99-9
C4H8O
72.11
WYURNTSHIVDZCO-UHFFFAOYSA-N
oxolane
C1CCOC1

T1368, 109-99-9, C4H8O

Spectrum™ Tetrahydrofuran, Anhydrous is used when performing certain reactions where the presence of water can prevent a chemical reaction from happening, or form undesirable products. Spectrum anhydrous chemical solvents are packaged in convenient sizes featuring a double closure, which includes an easy-slip-on vinyl cap with Teflon liner and screw-on phenolic cap and packaged in specially designed septum-sealed bottles under nitrogen to maintain low water content and ensure product integrity.

CAS Max % 1
Percent Purity 99.9%
Grade Reagent
Packaging Amber Glass Bottle
Residue after Evaporation 5e-006
Water 5e-005
Quantity 1 L