Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydrofuran, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
4L
Packaging:
Aluminum Bottle
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Bottle

Chemical Identifiers

109-99-9
C4H8O
72.107
WYURNTSHIVDZCO-UHFFFAOYSA-N
8028
CHEBI:26911
oxolane
C1CCOC1

High quality chemicals for laboratory and specialized industrial use

Formula Weight 72.11
Packaging Aluminum Bottle
Specific Gravity 1L = 0.88kg
Flash Point −14.00°C
Quantity 4L
Chemical Name or Material Tetrahydrofuran
Grade Baker Analyzed™ ACS Reagent