Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydrofuran, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
4L
500mL
Packaging:
Aluminum Bottle
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Bottle

Chemical Identifiers

109-99-9
C4H8O
72.107
WYURNTSHIVDZCO-UHFFFAOYSA-N
8028
CHEBI:26911

High quality chemicals for laboratory and specialized industrial use

CAS 109-99-9
Molecular Formula C4H8O
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
SMILES C1CCOC1
IUPAC Name oxolane
Molecular Weight (g/mol) 72.107
PubChem CID 8028
ChEBI CHEBI:26911
Formula Weight 72.11
Packaging Narrow Mouth PVC Coated Amber Bottle
Specific Gravity 1L = 0.88kg
Flash Point −14.00°C
Quantity 4L
Chemical Name or Material Tetrahydrofuran
Grade Baker Analyzed™ ACS Reagent
Show More Show Less