Alfa Aesar™ Tetrahydrofuran, Biograde, 99.8%, unstab.

Nucleic acid and peptide synthesis

Manufacturer: Alfa Aesar™ 44505M1

Quantity 4 x 1L

 View more versions of this product

Catalog No. AA44505M1

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 109-99-9
    Molecular Formula C4H8O
    Formula Weight 72.11
    MDL Number MFCD00005356
    UN Number UN2056
    Beilstein 102391
    Merck Index 14,9211
    Chemical Name or Material Tetrahydrofuran, Unstabilized
    Grade Biograde

    Specifications

    Formula Weight 72.11
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Flash Point −17°C (1°F)
    Boiling Point 66°C
    Physical Form Liquid
    Melting Point −108°C
    CAS 109-99-9
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: TETRAHYDROFURAN
    Color Colorless
    Beilstein 102391
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H351-H319-H335
    Highly flammable liquid and vapor.
    Suspected of causing cancer.
    Causes serious eye irritation.
    May cause respiratory irritation.
    RTECS Number LU5950000
    UN Number UN2056
    Merck Index 14,9211
    TSCA TSCA
    Odor Ether-like
    Refractive Index 1.4070
    Quantity 4 x 1L
    Molecular Formula C4H8O
    MDL Number MFCD00005356
    Density 0.889g/mL
    Chemical Name or Material Tetrahydrofuran, Unstabilized
    Assay 99.8%
    Grade Biograde

    GHS H Statement
    H225-H351-H319-H335
    Highly flammable liquid and vapor.
    Suspected of causing cancer.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit