Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Tetrahydrofuran-d8, 100%(Isotopic)

Manufacturer:  Alfa Aesar™ 42348SA

Quantity 2 each

 View more versions of this product

Catalog No. AA42348SA

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

1693-74-9
MFCD00044238
Tetrahydrofuran-d8, Isotopic
80.16
UN2056

Specifications

1693-74-9
0.985g/mL
−106°C
2 each
−17°C (−1°F)
MFCD00044238
Tetrahydrofuran-d8, Isotopic
Prescored Ampules
66°C
Liquid
Colorless
80.16
UN2056
100%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H302-H319-H335
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: TETRAHYDROFURAN

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit