Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydrofuran-D8, for NMR, 99.5 atom % D, ACROS Organics™

Click to view available options
Quantity:
1mL
5mL
10mL
25 mL
Packaging:
Glass Bottle
1693-74-9
C4H8O
80.16
MFCD00044238
WYURNTSHIVDZCO-SVYQBANQSA-N
tetrahydrofuran-d8, deuterated thf, octadeuterotetrahydrofuran, deuterated tetrahydrofuran, tetrahydrofuran-d8 thf-d8, tetrahydrofuran-d8, >=99.5 atom % d, 2h4 tetrahydro 2h4 furan, thf-d8, 2h8 tetrahydrofuran, furan-d4-, tetrahydro-d4
80290
2,2,3,3,4,4,5,5-octadeuteriooxolane
[2H]C1([2H])OC([2H])([2H])C([2H])([2H])C1([2H])[2H]
1693-74-9
C4H8O
80.16
MFCD00044238
WYURNTSHIVDZCO-SVYQBANQSA-N
tetrahydrofuran-d8, deuterated thf, octadeuterotetrahydrofuran, deuterated tetrahydrofuran, tetrahydrofuran-d8 thf-d8, tetrahydrofuran-d8, >=99.5 atom % d, 2h4 tetrahydro 2h4 furan, thf-d8, 2h8 tetrahydrofuran, furan-d4-, tetrahydro-d4
80290
2,2,3,3,4,4,5,5-octadeuteriooxolane
[2H]C1([2H])OC([2H])([2H])C([2H])([2H])C1([2H])[2H]
Color Colorless
Deuterium (D) 99.4% min.
Water 0.06% max. (Coulometric)
Quantity 1mL
Density 0.9800g/mL
Packaging Glass Bottle
Formula Weight 80.17
Physical Form Liquid
Grade NMR
Chemical Name or Material Tetrahydrofuran-D8, 99.5 atom % D

GHS H Statement:
Highly flammable liquid and vapour.
Causes serious eye irritation.
May cause respiratory irritation.
May form explosive peroxides.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 216-898-4

TSCA : TSCA