Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydrofuran, non-UV, HPLC Grade, 99.7+%, stab. with 250ppm BHT, Thermo Scientific Chemicals

Catalog Number AA38994K7
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
This item is not returnable. View return policy
109-99-9
C4H8O
72.11
MFCD00005356
WYURNTSHIVDZCO-UHFFFAOYSA-N
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
8028
CHEBI:26911
oxolane
C1CCOC1
This item is not returnable. View return policy
109-99-9
C4H8O
72.11
MFCD00005356
WYURNTSHIVDZCO-UHFFFAOYSA-N
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
8028
CHEBI:26911
oxolane
C1CCOC1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point °C to 108°C
Color Colorless
Appearance Colorless liquid
Density 0.889 g/cm3 at 20°C
Boiling Point 66°C
Quantity 4 L
Flash Point -17°C (1°F)
Odor Ether-like
Physical Form Liquid
Refractive Index 1.407
Assay Percent Range >99.7%
Vapor Pressure 200 hPa (150mm Hg) at 20°C
Beilstein 102391
Solubility Information Miscible with water.
Formula Weight 72.11
Grade HPLC
Chemical Name or Material Tetrahydrofuran, With 250ppm BHT
Show More Show Less
Hazard Category H225-H302-H319-H335-H336-H351
Hazard Statement Keep away from food, drink and animal feeding stuffs. Keep away from sources of ignition - No Smoking. Do not empty into drains. Take precautionary measures against static discharges. Wear suitable protective clothing and gloves. If swallowed seek medical advice immediately and show this container or label.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P330-P370+P378q-P501c
DOTInformation DOT Class: 3, Packaging Group: II
EINECSNumber 203-726-8
RTECSNumber LU5950000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit