Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydrofuran, Packaged under Argon, HPLC Grade, J.T.Baker™

Click to view available options
Quantity:
1L
4L
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Glass Bottle
109-99-9
C4H8O
72.107
WYURNTSHIVDZCO-UHFFFAOYSA-N
tetrahydrofuran,furan, tetrahydro,butylene oxide,furanidine,hydrofuran,tetramethylene oxide,oxacyclopentane,1,4-epoxybutane,tetrahydrofuranne,tetraidrofurano
8028
CHEBI:26911
oxolane
C1CCOC1
109-99-9
C4H8O
72.107
WYURNTSHIVDZCO-UHFFFAOYSA-N
tetrahydrofuran,furan, tetrahydro,butylene oxide,furanidine,hydrofuran,tetramethylene oxide,oxacyclopentane,1,4-epoxybutane,tetrahydrofuranne,tetraidrofurano
8028
CHEBI:26911
oxolane
C1CCOC1

High purity solvents for use in analytical and preparative separations

    For use in liquid chromatography (HPLC & UHPLC) & spectrophotometry
Formula Weight 72.11
Grade HPLC
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.88kg
Flash Point −14.00°C
Quantity 1L
Chemical Name or Material Tetrahydrofuran, Packaged under Argon