Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydrofuran, Ultra Low water, BakerDRY™, J.T.Baker™

Click to view available options
Quantity:
100mL
1L
Packaging:
Glass Bottle
Narrow Mouth Amber Glass Bottle, w/ Septum
109-99-9
C4H8O
72.107
WYURNTSHIVDZCO-UHFFFAOYSA-N
tetrahydrofuran,furan, tetrahydro,butylene oxide,furanidine,hydrofuran,tetramethylene oxide,oxacyclopentane,1,4-epoxybutane,tetrahydrofuranne,tetraidrofurano
8028
CHEBI:26911
oxolane
C1CCOC1
109-99-9
C4H8O
72.107
WYURNTSHIVDZCO-UHFFFAOYSA-N
tetrahydrofuran,furan, tetrahydro,butylene oxide,furanidine,hydrofuran,tetramethylene oxide,oxacyclopentane,1,4-epoxybutane,tetrahydrofuranne,tetraidrofurano
8028
CHEBI:26911
oxolane
C1CCOC1

Specifically manufactured for use in organometallic and oligonucleotide synthesis

    For Use in Organic Synthesis, including Organometallic Synthesis
Formula Weight 72.11
Grade BakerDRY™
Packaging Glass Bottle
Specific Gravity 1L = 0.88kg
Flash Point −14.00°C
Quantity 100mL
Chemical Name or Material Tetrahydrofuran, Ultra Low water