Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Tetrahydrofurfuryl acetate, 98%

Click to view available options
Quantity:
50g
250g
637-64-9
C7H12O3
144.17
MFCD00022478
AAQDYYFAFXGBFZ-UHFFFAOYSA-N
tetrahydrofurfuryl acetate,2-acetoxymethyloxolane,tetrahydrofuran-2-yl methyl acetate,2-furanmethanol, tetrahydro-, acetate,tetrahydro-2-furanmethyl acetate,furfuryl alcohol, tetrahydro-, acetate,2-acetoxymethyl tetrahydrofuran,tetrahydro-2-furanmethanol acetate,fema no. 3055,2-furanmethanol, tetrahydro-, 2-acetate
12506
oxolan-2-ylmethyl acetate
CC(=O)OCC1CCCO1
637-64-9
C7H12O3
144.17
MFCD00022478
AAQDYYFAFXGBFZ-UHFFFAOYSA-N
tetrahydrofurfuryl acetate,2-acetoxymethyloxolane,tetrahydrofuran-2-yl methyl acetate,2-furanmethanol, tetrahydro-, acetate,tetrahydro-2-furanmethyl acetate,furfuryl alcohol, tetrahydro-, acetate,2-acetoxymethyl tetrahydrofuran,tetrahydro-2-furanmethanol acetate,fema no. 3055,2-furanmethanol, tetrahydro-, 2-acetate
12506
oxolan-2-ylmethyl acetate
CC(=O)OCC1CCCO1
Flash Point 81°C (177°F)
Boiling Point 193°C to 195°C
Quantity 50g
Beilstein 383629
Formula Weight 144.17
Percent Purity 98%
Chemical Name or Material Tetrahydrofurfuryl acetate

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA