Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydrofurfuryl Alcohol 98%, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
Packaging:
Glass bottle
97-99-4
C5H10O2
102.13
MFCD00005372
BSYVTEYKTMYBMK-UHFFFAOYSA-N
tetrahydrofurfuryl alcohol,tetrahydro-2-furanmethanol,tetrahydrofuran-2-yl methanol,thfa,2-furanmethanol, tetrahydro,tetrahydro-2-furanylmethanol,qo thfa,tetrahydro-2-furancarbinol,tetrahydrofuryl carbinol,furfuryl alcohol, tetrahydro
7360
oxolan-2-ylmethanol
C1CC(OC1)CO
97-99-4
C5H10O2
102.13
MFCD00005372
BSYVTEYKTMYBMK-UHFFFAOYSA-N
tetrahydrofurfuryl alcohol,tetrahydro-2-furanmethanol,tetrahydrofuran-2-yl methanol,thfa,2-furanmethanol, tetrahydro,tetrahydro-2-furanylmethanol,qo thfa,tetrahydro-2-furancarbinol,tetrahydrofuryl carbinol,furfuryl alcohol, tetrahydro
7360
oxolan-2-ylmethanol
C1CC(OC1)CO
Viscosity 6.24 mPa.s (20°C)
Water 0.3% max.
Color Undesignated
Specific Gravity 1.0543
Refractive Index 1.4510 to 1.4530
Infrared Spectrum Authentic
Density 1.0543g/mL
Flash Point 74°C
Melting Point -80.0°C
Packaging Glass bottle
Quantity 1L
Boiling Point 178.0°C
Assay Percent Range 98%
Beilstein 17, V,3, 115
Merck Index 15, 9358
Solubility Information Solubility in water: soluble. Other solubilities: miscible with alcohol, ether, acetone, chloroform, and benzene
Formula Weight 102.13
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Tetrahydrofurfuryl alcohol, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.

GHS P Statement:
Wash face, hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

HYGROSCOPIC

EINECSNumber : 202-625-6

RTECSNumber : LU2450000

TSCA : TSCA