Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydrofurfuryl alcohol, 99%, Acros Organics

Click to view available options
Quantity:
1L
Packaging:
Glass bottle
97-99-4
C5H10O2
102.133
MFCD00005372
BSYVTEYKTMYBMK-UHFFFAOYSA-N
tetrahydrofurfuryl alcohol,tetrahydro-2-furanmethanol,tetrahydrofuran-2-yl methanol,thfa,2-furanmethanol, tetrahydro,tetrahydro-2-furanylmethanol,qo thfa,tetrahydro-2-furancarbinol,tetrahydrofuryl carbinol,furfuryl alcohol, tetrahydro
7360
oxolan-2-ylmethanol
C1CC(OC1)CO
97-99-4
C5H10O2
102.133
MFCD00005372
BSYVTEYKTMYBMK-UHFFFAOYSA-N
tetrahydrofurfuryl alcohol,tetrahydro-2-furanmethanol,tetrahydrofuran-2-yl methanol,thfa,2-furanmethanol, tetrahydro,tetrahydro-2-furanylmethanol,qo thfa,tetrahydro-2-furancarbinol,tetrahydrofuryl carbinol,furfuryl alcohol, tetrahydro
7360
oxolan-2-ylmethanol
C1CC(OC1)CO
Packaging Glass bottle
Color Undesignated
Water 0.3% max.
Refractive Index 1.4510 to 1.4530 (20°C, 589nm)
Density 1.0543g/mL
Infrared Spectrum Authentic
Quantity 1L
Assay Percent Range 99%
Beilstein 17, V,3, 115
Merck Index 15, 9358
Formula Weight 102.13
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material Tetrahydrofurfuryl alcohol, p.a.
Show More Show Less

GHS H Statement:
Causes serious eye irritation.

GHS P Statement:
Wash face, hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

HYGROSCOPIC