Alfa Aesar™ Tetrahydrofurfuryl chloride, 98%

Manufacturer: Alfa Aesar™ B2401522

Quantity 100g

 View more versions of this product

Catalog No. AAB2401522

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3003-84-7
    Molecular Formula C5H9ClO
    Formula Weight 120.58
    MDL Number MFCD00005370
    UN Number UN1993
    Beilstein 102716
    Synonym 2-(Chloromethyl)tetrahydrofuran
    Chemical Name or Material Tetrahydrofurfuryl chloride

    Specifications

    Formula Weight 120.58
    MDL Number MFCD00005370
    CAS 3003-84-7
    UN Number UN1993
    Synonym 2-(Chloromethyl)tetrahydrofuran
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapour.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 150° to 151°C
    Flash Point 47°C (116°F)
    Quantity 100g
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    TSCA TSCA
    Molecular Formula C5H9ClO
    Beilstein 102716
    RTECS Number LU0350000
    Chemical Name or Material Tetrahydrofurfuryl chloride
    Assay 98%

    Description

    Applications
    Tetrahydrofurfuryl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

    Solubility
    Not miscible or difficult to mix.

    Notes
    Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and alkali metals. It is sensitive to moisture.

    GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapour.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit