Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydropyran-2-methanol, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
100-72-1
C6H12O2
116.16
MFCD00006624
ROTONRWJLXYJBD-UHFFFAOYSA-N
tetrahydropyran-2-methanol,2h-pyran-2-methanol, tetrahydro,2-hydroxymethyltetrahydropyran,tetrahydropyran-2-carbinol,2-hydroxymethyl tetrahydropyran,pyran-2-methanol, tetrahydro,tetrahydro-2h-pyran-2-yl methanol,2-tetrahydropyranilcarbinol,tetrahydropyranyl-2-methanol,2-methyloltetrahydro-1,4-pyran
7524
oxan-2-ylmethanol
C1CCOC(C1)CO
100-72-1
C6H12O2
116.16
MFCD00006624
ROTONRWJLXYJBD-UHFFFAOYSA-N
tetrahydropyran-2-methanol,2h-pyran-2-methanol, tetrahydro,2-hydroxymethyltetrahydropyran,tetrahydropyran-2-carbinol,2-hydroxymethyl tetrahydropyran,pyran-2-methanol, tetrahydro,tetrahydro-2h-pyran-2-yl methanol,2-tetrahydropyranilcarbinol,tetrahydropyranyl-2-methanol,2-methyloltetrahydro-1,4-pyran
7524
oxan-2-ylmethanol
C1CCOC(C1)CO
Density 1.0200g/mL
Boiling Point 187°C
Flash Point 93°C
Refractive Index 1.4570 to 1.459
Specific Gravity 1.02
Quantity 10g
Packaging Glass bottle
Assay Percent Range 98%
Formula Weight 116.16
Chemical Name or Material Tetrahydropyran-2-methanol, 98%

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.