Alfa Aesar™ Tetrahydrothiophen-3-one, 97%

Manufacturer: Alfa Aesar™ A1100230

Quantity 250g

 View more versions of this product

Catalog No. AAA1100230

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1003-04-9
    Molecular Formula C4H6OS
    Formula Weight 102.16
    MDL Number MFCD00005412
    Beilstein 105201
    Synonym 4,5-Dihydro-3(2H)-thiophenone
    Chemical Name or Material Tetrahydrothiophen-3-one

    Specifications

    Formula Weight 102.16
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Synonym 4,5-Dihydro-3(2H)-thiophenone
    Health Hazard 2 GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    Boiling Point 174° to 175°C
    MDL Number MFCD00005412
    CAS 1003-04-9
    Health Hazard 1 Warning
    Flash Point 77°C (170°F)
    Odor Characteristic
    TSCA TSCA
    Molecular Formula C4H6OS
    Beilstein 105201
    Quantity 250g
    Chemical Name or Material Tetrahydrothiophen-3-one
    Assay 97%

    GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit