Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraisobutyldialuminoxane, 10 wt.% solution in toluene, AcroSeal™, Acros Organics

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass Bottle
998-00-5,108-88-3
C16H36Al2O
298.42
CTNFGBKAHAEKFE-UHFFFAOYSA-N
aluminum, tetrakis 2-methylpropyl-.mu.-oxodi,tetrakis isobutyl-mu-oxodialuminium,tetraisobutyl dialumoxane,aluminum, tetrakis 2-methylpropyl-mu-oxodi,bis diisobutylaluminum oxide,bis-diisobutylaluminum-oxide,tetraisobutyldialuminoxane solution,tetraisobutyldialuminoxane solution, 10 wt. % in toluene,bis 2-methylpropyl alumanyl oxy bis 2-methylpropyl alumane
16684086
bis(2-methylpropyl)alumanyloxy-bis(2-methylpropyl)alumane
CC(C)C[Al](CC(C)C)O[Al](CC(C)C)CC(C)C
998-00-5,108-88-3
C16H36Al2O
298.42
CTNFGBKAHAEKFE-UHFFFAOYSA-N
aluminum, tetrakis 2-methylpropyl-.mu.-oxodi,tetrakis isobutyl-mu-oxodialuminium,tetraisobutyl dialumoxane,aluminum, tetrakis 2-methylpropyl-mu-oxodi,bis diisobutylaluminum oxide,bis-diisobutylaluminum-oxide,tetraisobutyldialuminoxane solution,tetraisobutyldialuminoxane solution, 10 wt. % in toluene,bis 2-methylpropyl alumanyl oxy bis 2-methylpropyl alumane
16684086
bis(2-methylpropyl)alumanyloxy-bis(2-methylpropyl)alumane
CC(C)C[Al](CC(C)C)O[Al](CC(C)C)CC(C)C
Lewis acid
Boiling Point 111°C
Specific Gravity 0.867
Flash Point -8°C
Density 0.8670g/mL
Color Colorless
Quantity 100mL
Packaging AcroSeal™ Glass Bottle
Formula Weight 298.42
Physical Form Solution
Chemical Name or Material Tetraisobutyldialuminoxane

GHS H Statement
Highly flammable liquid and vapor.
Suspected of damaging the unborn child.
May be fatal if swallowed and enters airways.
May cause damage to organs through prolonged or repeated exposure.
May cause drowsines

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: Immediately call a POISON CENTER o

GHS Signal Word: Danger

EINECSNumber : 213-646-5