Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Tetraisopropyltin, 97+%

Manufacturer:  Alfa Aesar™ 3960909

Quantity 10g

 View more versions of this product

Catalog No. AA3960909


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

2949-42-0
291.05
UN2788
Sn[(CH3)2CH]4
MFCD00026346
Tetraisopropyltin

Specifications

112° to 114°C (18mmHg)
10g
1.124g/mL
1.4851
Liquid
291.05
UN2788
≥97%
2949-42-0
Characteristic
Colorless
Air & Moisture Sensitive
Sn[(CH3)2CH]4
MFCD00026346
Tetraisopropyltin

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.

RTECSNumber : WH8628000

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit